Learn More
MSC VENTURI PNEUM AIR BLOWER
Supplier:  MSC 03884251
Encompass
Venturi Style Pneumatic Blowers; SCFM @ 40 PSIG (Air Consumed): 35; SCFM @ 60 PSIG (Air Consumed): 45; SCFM @ 80 PSIG (Air Consumed): 62; Horn Material: Galvanized Steel; NPT Size (Inch): 1/2; Overall Length: 32.12 in; Face Diameter: 7.37 in; Base Slot Di
Catalog No. 50-273-9259