Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(2R)-1-methylsulfonylpropan-2-yl]-4-(trifluoromethyl)indole-5-carbonitrile | 1539314-06-1 | MFCD30343844 | 99.4% | 330.33 g·mol⁻¹ | C14H13F3N2O2S | 100 MG
SDP

Catalog No. 5000215582
Encompass_Preferred

GSK-2881078 is a nonsteroidal, orally active selective androgen receptor modulator (SARM) provided for research use in studies of muscle wasting, cachexia, and androgen receptor pharmacology. Supplied as a high-purity solid with recommended storage conditions, the compound is available in multiple sizes and in solution formats for convenient dosing and assay preparation.

  • High purity (99.4%) suitable for in vitro and in vivo studies.
  • White to off-white solid physical form.
  • Molecular weight 330.33 g·mol⁻¹.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Available in multiple sizes and DMSO solution formats for flexibility.

Catalog No. 50-002-15582 Supplier Medchemexpress LLC Supplier No. HY100186100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK-2881078 is a nonsteroidal, orally active selective androgen receptor modulator (SARM) provided for research use in studies of muscle wasting, cachexia, and androgen receptor pharmacology. Supplied as a high-purity solid with recommended storage conditions, the compound is available in multiple sizes and in solution formats for convenient dosing and assay preparation.

  • High purity (99.4%) suitable for in vitro and in vivo studies.
  • White to off-white solid physical form.
  • Molecular weight 330.33 g·mol⁻¹.
  • Recommended storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Available in multiple sizes and DMSO solution formats for flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.