Learn More
Medchemexpress LLC 1-[(2R)-1-methylsulfonylpropan-2-yl]-4-(trifluoromethyl)indole-5-carbonitrile | 1539314-06-1 | MFCD30343844 | 99.4% | 330.33 g/mol | C14H13F3N2O2S | 25 MG
SDP
Catalog No. 5000215869
Encompass_Preferred
GSK-2881078 is an orally active, nonsteroidal selective androgen receptor modulator used in preclinical research on muscle weakness and cachexia. It functions as a partial AR agonist and produces AR-mediated transcriptional activation in cellular assays. The compound is supplied as a characterized powder with established purity and storage recommendations for laboratory use.
  • Orally active nonsteroidal selective androgen receptor modulator.
  • Demonstrated AR-mediated transcriptional activation in cell assays.
  • High purity (99.42%) suitable for research applications.
  • Solid white to off-white powder form for easy handling.
  • Defined storage conditions to preserve stability.
  • Available in multiple package sizes for experimental flexibility.
Catalog No. 50-002-15869 Supplier Medchemexpress LLC Supplier No. HY10018625MG
GSK-2881078 is an orally active, nonsteroidal selective androgen receptor modulator used in preclinical research on muscle weakness and cachexia. It functions as a partial AR agonist and produces AR-mediated transcriptional activation in cellular assays. The compound is supplied as a characterized powder with established purity and storage recommendations for laboratory use.
  • Orally active nonsteroidal selective androgen receptor modulator.
  • Demonstrated AR-mediated transcriptional activation in cell assays.
  • High purity (99.42%) suitable for research applications.
  • Solid white to off-white powder form for easy handling.
  • Defined storage conditions to preserve stability.
  • Available in multiple package sizes for experimental flexibility.