Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(2R,3S,4R,5R)-3-fluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione | 69123-98-4 | MFCD00866922 | 99.9% | 372.09 | C9H10FIN2O5 | 1 ML
SDP

Catalog No. 5000226830
Encompass_Preferred

Fialuridine (FIAU) is a fluorinated, iodinated nucleoside analog with demonstrated antiviral activity supplied for laboratory research. The product is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for use in synthesis and biological testing.

  • Antiviral nucleoside analog active against hepatitis B virus and herpesviruses.
  • Supplied as 10 mM solution in DMSO (1 mL) for immediate use in assays.
  • Also available in multiple solid sizes for formulation or synthesis.
  • High reported purity (≈99.9%) suitable for research applications.
  • Molecular formula C9H10FIN2O5; molecular weight 372.09 Da.

Catalog No. 50-002-26830 Supplier Medchemexpress LLC Supplier No. HY11812210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fialuridine (FIAU) is a fluorinated, iodinated nucleoside analog with demonstrated antiviral activity supplied for laboratory research. The product is offered as a ready-to-use 10 mM solution in DMSO (1 mL) and as solid quantities for use in synthesis and biological testing.

  • Antiviral nucleoside analog active against hepatitis B virus and herpesviruses.
  • Supplied as 10 mM solution in DMSO (1 mL) for immediate use in assays.
  • Also available in multiple solid sizes for formulation or synthesis.
  • High reported purity (≈99.9%) suitable for research applications.
  • Molecular formula C9H10FIN2O5; molecular weight 372.09 Da.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.