Learn More
Ace Glass, Inc. PTFE 1/2in tube compression fitting to 1/2in male NPT
SDP
Catalog No. 50977685
Encompass
1/2 PTFE tube compression fitting to 1/2 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-685 Supplier Ace Glass, Inc. Supplier No. 1270948
1/2 PTFE tube compression fitting to 1/2 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.