Learn More
MSC Merit Brass K612-0806 1/2 x 3/8" 316 Stainless Steel Pipe Reducer Coupling FNPT x FNPT Ends, 150 psi
Catalog No. NC1631421
Encompass
Merit Brass K612-0806 1/2 x 3/8" 316 Stainless Steel Pipe Reducer Coupling FNPT x FNPT Ends, 150 psi
Catalog No. NC1631421 Supplier MSC Supplier No. 36901692
Merit Brass K612-0806 1/2 x 3/8" 316 Stainless Steel Pipe Reducer Coupling FNPT x FNPT Ends, 150 psi