Learn More
Medchemexpress LLC 1,3-17:0 DG-d5 | 1246523-65-8 | 99.9% | 602.00 | 1 MG
SDP
Catalog No. 5000363634
Encompass_Preferred
1,3-17:0 DG-d5 is the deuterium-labeled form of 1,3-17:0 DG. It is used as a tracer and as an internal standard in NMR, GC-MS, or LC-MS analysis for quantitative analysis. Deuteration has been shown to potentially impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled form of 1,3-17:0 DG
  • Used as a tracer
  • Used as an internal standard in NMR, GC-MS, or LC-MS analysis for quantitative analysis
  • Deuteration can potentially impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-63634 Supplier Medchemexpress LLC Supplier No. HY146832S1MG
1,3-17:0 DG-d5 is the deuterium-labeled form of 1,3-17:0 DG. It is used as a tracer and as an internal standard in NMR, GC-MS, or LC-MS analysis for quantitative analysis. Deuteration has been shown to potentially impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium-labeled form of 1,3-17:0 DG
  • Used as a tracer
  • Used as an internal standard in NMR, GC-MS, or LC-MS analysis for quantitative analysis
  • Deuteration can potentially impact pharmacokinetic and metabolic profiles of drugs