Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,2-Dioxathiolane 2,2-dioxide, 96%, Thermo Scientific Chemicals

Catalog No. AC426630050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC426630050 5 g Glass bottle
AC426630010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC426630050 Supplier Thermo Scientific Chemicals Supplier No. 426630050
  • $121.00
    $101.65 / Each of 1

    Save $19.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1072-53-3 | Molecular Formula: C2H4O4S | Molecular Weight: 124.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-53-3
Molecular Formula C2H4O4S
Molecular Weight (g/mol) 124.12
MDL Number MFCD00221769
InChI Key ZPFAVCIQZKRBGF-UHFFFAOYSA-N
Synonym ethylenesulfate, ethylene sulfate, ethylene glycol, cyclic sulfate, glycol sulfate, 1,2-ethylene sulfate, 1,3,2-dioxathiolane, 2,2-dioxide, sulfuric acid, cyclic ethylene ester, 1,3,2-dioxathiolane-2,2-dioxide, 1,3,2??-dioxathiolane-2,2-dione, ethosulfate
PubChem CID 14075
IUPAC Name 1,3,2-dioxathiolane 2,2-dioxide
SMILES C1COS(=O)(=O)O1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 95°C to 97°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 124.12
Percent Purity 96%
Chemical Name or Material 1,3,2-Dioxathiolane 2,2-dioxide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.