Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[3-[3-(4-chlorophenyl)propoxy]propyl]piperidine | 362665-56-3 | 100 MG
SDP

Catalog No. 5000230535
Encompass_Preferred

Pitolisant is a selective histamine H3 receptor inverse agonist supplied as a research standard for receptor pharmacology and analytical applications. It is intended for use in binding assays, pharmacology studies, and analytical method development.

  • Selective H3 receptor inverse agonist (Ki = 0.16 nM).
  • Used as an analytical reference material and in pharmacology studies.
  • Chemical formula C17H26ClNO; molecular weight 295.8 g·mol-1.
  • CAS number 362665-56-3; MFCD18782650.
  • Available as a 100 mg research standard; handle per laboratory safety procedures.

Catalog No. 50-002-30535 Supplier Medchemexpress LLC Supplier No. HY12199100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitolisant is a selective histamine H3 receptor inverse agonist supplied as a research standard for receptor pharmacology and analytical applications. It is intended for use in binding assays, pharmacology studies, and analytical method development.

  • Selective H3 receptor inverse agonist (Ki = 0.16 nM).
  • Used as an analytical reference material and in pharmacology studies.
  • Chemical formula C17H26ClNO; molecular weight 295.8 g·mol-1.
  • CAS number 362665-56-3; MFCD18782650.
  • Available as a 100 mg research standard; handle per laboratory safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.