Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[(3,3-Diphenylpropyl)(methyl)amino]-2-methyl-2-propanol 98.0+%, TCI America™
SDP

Catalog No. D471325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D471325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D471325G Supplier TCI America Supplier No. D471325G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 100442-33-9
Molecular Formula C20H27NO
Molecular Weight (g/mol) 297.442
MDL Number MFCD11100703
InChI Key MQWDISMNBYOLAB-UHFFFAOYSA-N
PubChem CID 10851563
IUPAC Name 1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-ol
SMILES CC(C)(CN(C)CCC(C1=CC=CC=C1)C2=CC=CC=C2)O

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 g
Formula Weight 297.44
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-[(3,3-Diphenylpropyl)(methyl)amino]-2-methyl-2-propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.