Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,3,3-Trimethyl-2-methyleneindoline

Catalog No. M46209100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M46209100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M46209100G Supplier Sigma Aldrich Supplier No. M46209100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,3,3-Trimethyl-2-methyleneindoline

Specifications

CAS 118-12-7
Density 0.979 g/mL (at 25°C (literature))
Boiling Point 248°C (lit.)
Molecular Formula C12H15N
Refractive Index n20/D 1.577 (literature)
Linear Formula C12H15N
MDL Number MFCD00005813
Quantity 5 g
Synonym 2-Methylene-1,3,3-trimethylindoline; Fischer base; NSC 66176
Molecular Weight (g/mol) 173.25
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.