Learn More
Medchemexpress LLC 1,3,4,5-tetrahydro-4-methyl-6-[1-methyl-3-(1-methyl-pyrazol-4-yl)-indazol-5-yl]-benzodiazepin-2-one | 1884712-47-3 | 100.0% | 386.45 g/mol | C22H22N6O | 1 ML
SDP
Catalog No. 5000215451
Encompass_Preferred
CPI-637 is a selective, cell-active benzodiazepinone inhibitor of CBP and EP300 bromodomains with potent biochemical activity and demonstrated cellular effects. It inhibits CBP and EP300 with low-nanomolar IC50 values, shows reduced activity against BRD4 BD-1, and is supplied as a concentrated solution for research use.
  • Selective CBP and EP300 bromodomain inhibition.
  • Potent biochemical activity (low-nanomolar IC50 values).
  • Cell-active with measurable EC50 in cellular assays.
  • Supplied as a concentrated solution for convenient dosing in assays.
  • Suitable for epigenetics and cancer biology research applications.
  • Reported high chemical purity for reproducible results.
Catalog No. 50-002-15451 Supplier Medchemexpress LLC Supplier No. HY10048210MM 1ML
CPI-637 is a selective, cell-active benzodiazepinone inhibitor of CBP and EP300 bromodomains with potent biochemical activity and demonstrated cellular effects. It inhibits CBP and EP300 with low-nanomolar IC50 values, shows reduced activity against BRD4 BD-1, and is supplied as a concentrated solution for research use.
  • Selective CBP and EP300 bromodomain inhibition.
  • Potent biochemical activity (low-nanomolar IC50 values).
  • Cell-active with measurable EC50 in cellular assays.
  • Supplied as a concentrated solution for convenient dosing in assays.
  • Suitable for epigenetics and cancer biology research applications.
  • Reported high chemical purity for reproducible results.