Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-beta-D-glucopyranose 97.0+%, TCI America™
SDP

Catalog No. T2196200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2196200MG 200 mg
T21961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2196200MG Supplier TCI America Supplier No. T2196200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80321-89-7
Molecular Formula C14H19N3O9
Molecular Weight (g/mol) 373.318
InChI Key QKGHBQJLEHAMKJ-DHGKCCLASA-N
PubChem CID 133445
IUPAC Name [(2R,3S,4R,5R,6S)-3,4,6-triacetyloxy-5-azidooxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)OC(=O)C)N=[N+]=[N-])OC(=O)C)OC(=O)C

Specifications

Melting Point 97°C
Color White
Quantity 200 mg
Formula Weight 373.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,4,6-Tetra-O-acetyl-2-azido-2-deoxy-beta-D-glucopyranose
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.