Learn More
Medchemexpress LLC 1,3,4,6-tetra-O-acetyl-N-azidoacetylmannosamine | 361154-30-5 | MFCD26961126 | 99.7% | 430.37 g/mol | C16H22N4O10 | 5 MG
SDP
Catalog No. 5000227693
Encompass_Preferred
Ac4ManNAz is a peracetylated, azido-functionalized monosaccharide used for metabolic glycoprotein labeling. It introduces an azide bioorthogonal handle into cellular glycans that can be targeted by click chemistry for visualization, enrichment, or proteomic analysis.
  • Copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) compatible.
  • Peracetylation improves cell permeability for efficient metabolic incorporation.
  • Suitable for cell-surface labeling, imaging, and proteomic enrichment workflows.
  • High reported purity (≈99.7%) supports reproducible experimental results.
  • Solid, white to light yellow appearance for straightforward handling.
  • Molecular weight 430.37 g/mol and formula C16H22N4O10 for exact mass calculations.
Catalog No. 50-002-27693 Supplier Medchemexpress LLC Supplier No. HY1182975MG
Ac4ManNAz is a peracetylated, azido-functionalized monosaccharide used for metabolic glycoprotein labeling. It introduces an azide bioorthogonal handle into cellular glycans that can be targeted by click chemistry for visualization, enrichment, or proteomic analysis.
  • Copper-catalyzed azide-alkyne cycloaddition (CuAAC) and strain-promoted azide-alkyne cycloaddition (SPAAC) compatible.
  • Peracetylation improves cell permeability for efficient metabolic incorporation.
  • Suitable for cell-surface labeling, imaging, and proteomic enrichment workflows.
  • High reported purity (≈99.7%) supports reproducible experimental results.
  • Solid, white to light yellow appearance for straightforward handling.
  • Molecular weight 430.37 g/mol and formula C16H22N4O10 for exact mass calculations.