Learn More
Cayman Chemical 1 3 4 6TetraOacetyl2azId 10mg
SDP
Catalog No. 502992230
Encompass_Preferred
A ManNAc analog and building block; has been used as a precursor in the synthesis of differently substituted trimers of the group A N. meningitidis capsular polysaccharide repeating unit for use in immunological experiments; has also been used in the synthesis of fluorescent probes for lysosomal labeling
Catalog No. 50-299-2230 Supplier Cayman Chemical Supplier No. 3054210 MG
A ManNAc analog and building block; has been used as a precursor in the synthesis of differently substituted trimers of the group A N. meningitidis capsular polysaccharide repeating unit for use in immunological experiments; has also been used in the synthesis of fluorescent probes for lysosomal labeling