Learn More
Medchemexpress LLC [1-(3,4-dichlorophenyl)-1H-pyrrolo[2,3-b]pyridin-2-yl](3,3-difluoro-1-azetidinyl)-methanone | 2410550-31-9 | 99.8% | C17H11Cl2F2N3O | 10MG
SDP
Catalog No. 5000260266
Encompass_Preferred
KVA-D-88 is a research-grade small-molecule inhibitor selective for phosphodiesterase 4B (PDE4B). It is brain-penetrant and has been used in preclinical studies to probe PDE4-mediated signaling and cocaine-related behavioral effects.
  • Selective PDE4B inhibition; reported IC50 140 nM (PDE4D IC50 880 nM).
  • Brain-penetrant small molecule suitable for in vivo studies.
  • High chemical purity (>99.8%).
  • Solid, white to off-white powder form.
  • Stable when stored at recommended temperatures for long-term use.
  • Supplied for research use only; not for human or veterinary use.
Catalog No. 50-002-60266 Supplier Medchemexpress LLC Supplier No. HY13917810MG
KVA-D-88 is a research-grade small-molecule inhibitor selective for phosphodiesterase 4B (PDE4B). It is brain-penetrant and has been used in preclinical studies to probe PDE4-mediated signaling and cocaine-related behavioral effects.
  • Selective PDE4B inhibition; reported IC50 140 nM (PDE4D IC50 880 nM).
  • Brain-penetrant small molecule suitable for in vivo studies.
  • High chemical purity (>99.8%).
  • Solid, white to off-white powder form.
  • Stable when stored at recommended temperatures for long-term use.
  • Supplied for research use only; not for human or veterinary use.