Learn More
Medchemexpress LLC (±)-1-(3,4-dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride | 112282-24-3 | MFCD00672679 | >98.0% | 391.76 g·mol⁻¹ | C18H25Cl3N2O | 50 MG
SDP
Catalog No. 5000211684
Encompass_Preferred
BRL 52537 hydrochloride is a selective kappa-opioid receptor (KOR) agonist used in preclinical research. It exhibits potent KOR activity and has been reported to reduce ischemia-evoked nitric oxide production and attenuate early stroke-related damage. The compound is supplied as a solid for laboratory use and should be stored sealed and protected from moisture at recommended temperatures.
  • Selective kappa-opioid receptor agonist.
  • Potent KOR affinity with low nanomolar activity.
  • Demonstrated neuroprotective effects in preclinical studies.
  • Suitable for in vitro and in vivo research applications.
  • Store sealed and protected from moisture under recommended conditions.
Catalog No. 50-002-11684 Supplier Medchemexpress LLC Supplier No. HY10107950MG
BRL 52537 hydrochloride is a selective kappa-opioid receptor (KOR) agonist used in preclinical research. It exhibits potent KOR activity and has been reported to reduce ischemia-evoked nitric oxide production and attenuate early stroke-related damage. The compound is supplied as a solid for laboratory use and should be stored sealed and protected from moisture at recommended temperatures.
  • Selective kappa-opioid receptor agonist.
  • Potent KOR affinity with low nanomolar activity.
  • Demonstrated neuroprotective effects in preclinical studies.
  • Suitable for in vitro and in vivo research applications.
  • Store sealed and protected from moisture under recommended conditions.