Learn More
Medchemexpress LLC (±)-1-(3,4-dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride | 112282-24-3 | MFCD00672679 | 98.1% | 391.76 g/mol | C18H25Cl3N2O | 10 MG
SDP
Catalog No. 5000211832
Encompass_Preferred
BRL 52537 hydrochloride is a potent, highly selective κ-opioid receptor (KOR) agonist provided as a solid for laboratory research. It is used in pharmacology and neuroscience to study KOR-mediated signaling and potential neuroprotective effects. Manufacturer documentation includes analytical data and handling instructions.
  • Potent KOR agonist with Ki = 0.24 nM.
  • High selectivity versus μ-opioid receptor (Ki ≈ 1,560 nM).
  • High purity (~98.1%) suitable for research purposes.
  • Off-white to light yellow solid, convenient for weighing.
  • Soluble in methanol (25 mg/mL) and DMSO (5 mg/mL with warming).
  • Store sealed at 4°C; in solution store at -80°C for long-term stability.
Catalog No. 50-002-11832 Supplier Medchemexpress LLC Supplier No. HY10107910MG
BRL 52537 hydrochloride is a potent, highly selective κ-opioid receptor (KOR) agonist provided as a solid for laboratory research. It is used in pharmacology and neuroscience to study KOR-mediated signaling and potential neuroprotective effects. Manufacturer documentation includes analytical data and handling instructions.
  • Potent KOR agonist with Ki = 0.24 nM.
  • High selectivity versus μ-opioid receptor (Ki ≈ 1,560 nM).
  • High purity (~98.1%) suitable for research purposes.
  • Off-white to light yellow solid, convenient for weighing.
  • Soluble in methanol (25 mg/mL) and DMSO (5 mg/mL with warming).
  • Store sealed at 4°C; in solution store at -80°C for long-term stability.