Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(3,4-dihydroxy-5-nitrophenyl)-2-phenylethanone | 274925-86-9 | MFCD09954136 | 99.9% | 273.24 | C14H11NO5 | 50 MG
SDP

Catalog No. 5000242826
Encompass_Preferred

Nebicapone is a reversible catechol-O-methyltransferase (COMT) inhibitor provided as a solid research compound for in vitro and in vivo studies. It is used to study COMT-mediated metabolism of L-DOPA and related pharmacology, and is supplied with characterization data and storage recommendations for experimental stability.

  • Reversible COMT inhibitor for research use.
  • Reported purity 99.9%.
  • Molecular weight 273.24 g/mol.
  • Physical form solid powder.
  • High solubility in DMSO (≥100 mg/mL) and formulated in vivo solubility ≥2.5 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline.
  • Recommended storage 4°C protected from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-42826 Supplier Medchemexpress LLC Supplier No. HY10640550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nebicapone is a reversible catechol-O-methyltransferase (COMT) inhibitor provided as a solid research compound for in vitro and in vivo studies. It is used to study COMT-mediated metabolism of L-DOPA and related pharmacology, and is supplied with characterization data and storage recommendations for experimental stability.

  • Reversible COMT inhibitor for research use.
  • Reported purity 99.9%.
  • Molecular weight 273.24 g/mol.
  • Physical form solid powder.
  • High solubility in DMSO (≥100 mg/mL) and formulated in vivo solubility ≥2.5 mg/mL in 10% DMSO/40% PEG300/5% Tween-80/45% saline.
  • Recommended storage 4°C protected from light; in solution store at -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.