Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(3,4-dimethoxyphenyl)-3-(3,4,5-trimethoxyphenyl)-2-propen-1-one | 111797-22-9 | MFCD01060969 | 99.9% | 358.39 | C20H22O6 | 25 MG
SDP

Catalog No. 5000248495
Encompass_Preferred

MD2-IN-1 is a small-molecule inhibitor of myeloid differentiation protein 2 (MD2) that binds recombinant human MD2 with a reported KD of 189 μM. Supplied for research use, it is intended for biochemical and cellular assays probing MD2/TLR signaling.

  • Reported KD 189 μM for recombinant human MD2.
  • Molecular formula C20H22O6; molecular weight 358.39.
  • CAS number 111797-22-9.
  • High purity (≈99.9%).
  • Available pack sizes include 25 MG solid and 10 mM solutions.
  • Storage: powder -20°C (long term) or 4°C; in solvent -80°C.

Catalog No. 50-002-48495 Supplier Medchemexpress LLC Supplier No. HY10348325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MD2-IN-1 is a small-molecule inhibitor of myeloid differentiation protein 2 (MD2) that binds recombinant human MD2 with a reported KD of 189 μM. Supplied for research use, it is intended for biochemical and cellular assays probing MD2/TLR signaling.

  • Reported KD 189 μM for recombinant human MD2.
  • Molecular formula C20H22O6; molecular weight 358.39.
  • CAS number 111797-22-9.
  • High purity (≈99.9%).
  • Available pack sizes include 25 MG solid and 10 mM solutions.
  • Storage: powder -20°C (long term) or 4°C; in solvent -80°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.