Learn More
Medchemexpress LLC 1-[(3,4-dimethoxyphenyl)methyl]-2,3,4,9-tetrahydro-6-methyl-1H-pyrido[3,4-b]indole hydrochloride | 172895-15-7 | 99.5% | 372.89 g·mol⁻¹ | C21H24N2O2·HCl | 100 MG
SDP
Catalog No. 5000211724
Encompass_Preferred
LY-272015 hydrochloride is a selective, orally active 5-HT2B receptor antagonist supplied as the hydrochloride salt for research use. Provided as an off-white to light yellow solid, it is used in pharmacology and receptor-binding studies to probe serotonergic signaling and cardiovascular effects in preclinical models.
  • Selective 5-HT2B receptor antagonist for targeted pharmacological studies.
  • High purity suitable for research and assay development.
  • Hydrochloride salt in solid form for stable handling and storage.
  • Applicable to in vitro receptor assays and in vivo pharmacology studies.
  • Well characterized in the literature with established physicochemical data.
Catalog No. 50-002-11724 Supplier Medchemexpress LLC Supplier No. HY100851A100MG
LY-272015 hydrochloride is a selective, orally active 5-HT2B receptor antagonist supplied as the hydrochloride salt for research use. Provided as an off-white to light yellow solid, it is used in pharmacology and receptor-binding studies to probe serotonergic signaling and cardiovascular effects in preclinical models.
  • Selective 5-HT2B receptor antagonist for targeted pharmacological studies.
  • High purity suitable for research and assay development.
  • Hydrochloride salt in solid form for stable handling and storage.
  • Applicable to in vitro receptor assays and in vivo pharmacology studies.
  • Well characterized in the literature with established physicochemical data.