Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3,4-Dimethylphenyl)hydrazine hydrochloride, 90%, Thermo Scientific™

Catalog No. BTB08676DA
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BTB08676DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB08676DA Supplier Thermo Scientific Chemicals Supplier No. BTB08676DA

missing translation for 'validMainframeMsg'

Specifications

CAS 60481-51-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C8H13ClN2
Quantity 1g
MDL Number MFCD00052270
Synonym 3,4-dimethylphenylhydrazine hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride, 1-3,4-dimethylphenyl hydrazine hydrochloride, hydrazine, 3,4-dimethylphenyl-, monohydrochloride, 4-hydrazino-o-xylene hydrochloride, 3,4-dimethylphenyl hydrazine hydrochloride 1:x, hydrazine, 3,4-dimethylphenyl-, hydrochloride, 3,4-dimethylphenyl diazane hydrochloride, pubchem7555, 3,4-dimethylphenylhydrazinehydrochloride
InChI Key YYMIOVAEQIEPET-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)NN)C.Cl
IUPAC Name (3,4-dimethylphenyl)hydrazine;hydrochloride
Molecular Weight (g/mol) 172.656
PubChem CID 173740
Formula Weight 172.66
Percent Purity 97%
Chemical Name or Material 1-(3, 4-Dimethylphenyl)hydrazine hydrochloride
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.