Learn More
Medchemexpress LLC 1,3,4-oxadiazole | 288-99-3 | MFCD00955535 | 99.8% | 70.05 g/mol | C2H2N2O | 100 MG
SDP
Catalog No. 5000283502
Encompass_Preferred
1,3,4-Oxadiazole is a five-membered heterocyclic compound used as a versatile building block and scaffold in medicinal chemistry and synthetic research. It is provided with analytical documentation for laboratory research use only and is intended for non-clinical applications.
  • High purity suitable for research applications.
  • Supplied with certificate of analysis and safety data sheet for traceability.
  • Commonly used as a synthetic building block and pharmacophore scaffold.
  • Small, planar heterocycle with formula C2H2N2O and molecular weight 70.05 g/mol.
  • Available in multiple package sizes, including small research quantities.
  • Colorless to light yellow liquid with typical density around 0.97 g/cm3.
Catalog No. 50-002-83502 Supplier Medchemexpress LLC Supplier No. HYW131122100MG
1,3,4-Oxadiazole is a five-membered heterocyclic compound used as a versatile building block and scaffold in medicinal chemistry and synthetic research. It is provided with analytical documentation for laboratory research use only and is intended for non-clinical applications.
  • High purity suitable for research applications.
  • Supplied with certificate of analysis and safety data sheet for traceability.
  • Commonly used as a synthetic building block and pharmacophore scaffold.
  • Small, planar heterocycle with formula C2H2N2O and molecular weight 70.05 g/mol.
  • Available in multiple package sizes, including small research quantities.
  • Colorless to light yellow liquid with typical density around 0.97 g/cm3.