Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3,4-oxadiazole | 288-99-3 | MFCD00955535 | 99.8% | 70.05 g/mol | C2H2N2O | 250 MG
SDP

Catalog No. 5000283501
Encompass_Preferred

1,3,4-Oxadiazole is a small heterocyclic compound commonly used as a building block and intermediate in medicinal chemistry and chemical research. The material is supplied as a high-purity research sample in milligram to gram pack sizes and is characterized by a low molecular weight and simple ring structure.

  • Purity 99.8%.
  • Molecular formula C2H2N2O; molecular weight 70.05 g/mol.
  • Appearance colorless to light yellow liquid (density ~0.97 g/cm3).
  • Suitable as a synthetic building block for medicinal chemistry applications.
  • Storage recommendations: pure form stable at -20°C for extended periods; shorter stability in solution.
  • Available in mg and g pack sizes, including 250 MG.

Catalog No. 50-002-83501 Supplier Medchemexpress LLC Supplier No. HYW131122250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3,4-Oxadiazole is a small heterocyclic compound commonly used as a building block and intermediate in medicinal chemistry and chemical research. The material is supplied as a high-purity research sample in milligram to gram pack sizes and is characterized by a low molecular weight and simple ring structure.

  • Purity 99.8%.
  • Molecular formula C2H2N2O; molecular weight 70.05 g/mol.
  • Appearance colorless to light yellow liquid (density ~0.97 g/cm3).
  • Suitable as a synthetic building block for medicinal chemistry applications.
  • Storage recommendations: pure form stable at -20°C for extended periods; shorter stability in solution.
  • Available in mg and g pack sizes, including 250 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.