Learn More
Medchemexpress LLC 1,3,4-oxadiazole | 288-99-3 | MFCD00955535 | 99.8% | 70.05 g·mol⁻¹ | C2H2N2O | 500 MG
SDP
Catalog No. 5000283367
Encompass_Preferred
1,3,4-Oxadiazole is a small heterocyclic compound used as a synthetic building block in medicinal chemistry and organic synthesis. It is provided at high purity for research use and is suitable as an intermediate or scaffold in small-molecule synthesis.
  • High purity for research applications (99.8%).
  • Colorless to light yellow liquid; density 0.97 g·cm⁻³.
  • Stable when stored under recommended conditions.
  • Useful scaffold for heterocycle synthesis and library design.
  • Available in multiple laboratory pack sizes for flexibility.
  • Supplied for research use only.
Catalog No. 50-002-83367 Supplier Medchemexpress LLC Supplier No. HYW131122500MG
1,3,4-Oxadiazole is a small heterocyclic compound used as a synthetic building block in medicinal chemistry and organic synthesis. It is provided at high purity for research use and is suitable as an intermediate or scaffold in small-molecule synthesis.
  • High purity for research applications (99.8%).
  • Colorless to light yellow liquid; density 0.97 g·cm⁻³.
  • Stable when stored under recommended conditions.
  • Useful scaffold for heterocycle synthesis and library design.
  • Available in multiple laboratory pack sizes for flexibility.
  • Supplied for research use only.