Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,3,4-thiadiazole-2,5-dithiol | 1072-71-5 | MFCD00003103 | 100.0% | 150.25 g/mol | C2H2N2S3 | 25 G
SDP

Catalog No. 5000302852
Encompass_Preferred

1,3,4-Thiadiazole-2,5-dithiol is an analytical reagent used for the determination of bismuth, copper, and lead. It is supplied as a high-purity solid suitable for biochemical assay and metal detection methods.

  • High purity (99.98%) for reliable analytical results.
  • Solid form, easy to weigh and handle.
  • Molecular weight 150.25 g/mol and formula C2H2N2S3 for reference.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Suitable for metal determination assays and biochemical reagent applications.

Catalog No. 50-003-02852 Supplier Medchemexpress LLC Supplier No. 25GHYW032388
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,3,4-Thiadiazole-2,5-dithiol is an analytical reagent used for the determination of bismuth, copper, and lead. It is supplied as a high-purity solid suitable for biochemical assay and metal detection methods.

  • High purity (99.98%) for reliable analytical results.
  • Solid form, easy to weigh and handle.
  • Molecular weight 150.25 g/mol and formula C2H2N2S3 for reference.
  • Soluble in DMSO for preparation of concentrated stock solutions.
  • Suitable for metal determination assays and biochemical reagent applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.