Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Adamantanetriol 97.0+%, TCI America™
SDP

Catalog No. A16825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16825G 5 g
A1682250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16825G Supplier TCI America Supplier No. A16825G

Chemical Identifiers

CAS 99181-50-7
Molecular Formula C10H16O3
Molecular Weight (g/mol) 184.235
InChI Key MCYBYTIPMYLHAK-UHFFFAOYSA-N
Synonym 1,3,5-Trihydroxyadamantane
PubChem CID 9899022
IUPAC Name adamantane-1,3,5-triol
SMILES C1C2CC3(CC1(CC(C2)(C3)O)O)O

Specifications

Color White
Quantity 5 g
Formula Weight 184.24
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Adamantanetriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.