Learn More
Medchemexpress LLC 1,3,5-Benzenetricarboxylic acid | 554-95-0 | 99.8% | C9H6O6 | 25 G
SDP
Catalog No. 5000357572
Encompass_Preferred
Benzene-1,3,5-tricarboxylic acid, also known as Trimesic acid, serves as a scaffold material. This compound has been shown to improve the binding capabilities of bicyclic peptides to proteins and to increase the mechanical strength of hydrogels.
  • Acts as a scaffold.
  • Enhances the binding ability of bicyclic peptides to proteins.
  • Improves the mechanical stability of hydrogels.
Catalog No. 50-003-57572 Supplier Medchemexpress LLC Supplier No. 25GHY78985
Benzene-1,3,5-tricarboxylic acid, also known as Trimesic acid, serves as a scaffold material. This compound has been shown to improve the binding capabilities of bicyclic peptides to proteins and to increase the mechanical strength of hydrogels.
  • Acts as a scaffold.
  • Enhances the binding ability of bicyclic peptides to proteins.
  • Improves the mechanical stability of hydrogels.