Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Benzenetricarboxylic acid, 98%, Thermo Scientific™

Catalog No. AC105351000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC105351000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC105351000 Supplier Thermo Scientific Chemicals Supplier No. 105351000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-95-0
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002517
InChI Key QMKYBPDZANOJGF-UHFFFAOYSA-N
Synonym trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
PubChem CID 11138
ChEBI CHEBI:46032
IUPAC Name benzene-1,3,5-tricarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

Melting Point 380.0°C
Flash Point >250°C
Linear Formula C6H3(CO2H)3
Quantity 100 g
Solubility Information Solubility in water: 2.6g/L (25°C) - 7.9g/L (50°C). Other solubilities: 1.24 g/100g ethyl ether - 8 g/100g methanol, 0.46 g/100g acetic acid - >20.00 g/100g thf (25°C), 1.88 g/100g acetone - <0.01 g/100g xylene (25°C), <0.01 g/100g ccl4 - <0.01 g/100g m
Formula Weight 210.14
Chemical Name or Material 1,3,5-Benzenetricarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.