Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Benzenetricarboxylic acid chloride, 98%

Catalog No. AC164375000
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164375000 500 g Glass bottle
AC164370050 5 g Glass bottle
AC164371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC164375000 Supplier Thermo Scientific Chemicals Supplier No. 164375000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4422-95-1 | C9H3Cl3O3 | 265.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4422-95-1
Molecular Formula C9H3Cl3O3
Molecular Weight (g/mol) 265.47
MDL Number MFCD00000679
InChI Key UWCPYKQBIPYOLX-UHFFFAOYSA-N
Synonym trimesoyl chloride, 1,3,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylic acid chloride, 1,3,5-benzenetricarbonyl chloride, trimesic acid trichloride, 1,3,5-benzenetricarboxylic chloride, benzene-1,3,5-tricarbonyl trichloride, unii-5866peo79w, 1,5-benzenetricarbonyl trichloride, 1,3,5-benzenetricarboxylicacidchloride
PubChem CID 78138
IUPAC Name benzene-1,3,5-tricarbonyl chloride
SMILES ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 34.5°C to 36°C
Color Colorless to Yellow
Boiling Point 180°C (16.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (ex Cl) (Argentometry)
Packaging Glass bottle
Refractive Index 1.5945 to 1.5965
Linear Formula C6H3(COCl)3
Quantity 500 g
Beilstein 09, IV, 3748
Solubility Information Solubility in water: decomposes
Formula Weight 265.47
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3, 5-Benzenetricarboxylic acid chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-594-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.