Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Benzenetricarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B0043100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0043100G 100 g
B004325G 25 g
B0043500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0043100G Supplier TCI America Supplier No. B0043100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 554-95-0
Molecular Formula C9H6O6
Molecular Weight (g/mol) 210.14
MDL Number MFCD00002517
InChI Key QMKYBPDZANOJGF-UHFFFAOYSA-N
Synonym trimesic acid, 1,3,5-benzenetricarboxylic acid, trimesinic acid, trimesitinic acid, 5-carboxyisophthalic acid, 1,3,5-benzene tricarboxylic acid, 1,3,5-tricarboxybenzene, unii-ou36oo5mtn, ou36oo5mtn, chembl77562
PubChem CID 11138
ChEBI CHEBI:46032
IUPAC Name benzene-1,3,5-tricarboxylic acid
SMILES OC(=O)C1=CC(=CC(=C1)C(O)=O)C(O)=O

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 210.14
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Benzenetricarboxylic Acid
EINECSNumber (3)-1279
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.