Learn More
Strem, An Ascensus Company CAS# 620960-26-1. 250mg. 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1R,2R)-(-)-2-(dimethylamino)cyclohexyl]thiourea (R,R-TUC). MFCD09834839
SDP
Supplier:  Strem, An Ascensus Company 070283250MG
Encompass
CAS# 620960-26-1. 250mg. 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1R,2R)-(-)-2-(dimethylamino)cyclohexyl]thiourea (R,R-TUC). MFCD09834839. Molecular Weight: 413.42. Molecular Formula: C17H21F6N3S. Color/form: white pwdr. Strem# 07-0283. http://www.strem.com/catalog/v/07-0283/
Catalog No. 50-901-08921