Learn More
Strem, An Ascensus Company CAS# 851477-20-8. 1g. 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea (S,S-TUC). MFCD09834840
SDP
Catalog No. 5090108924
Encompass
CAS# 851477-20-8. 1g. 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea (S,S-TUC). MFCD09834840. Molecular Weight: 413.42. Molecular Formula: C17H21F6N3S. Color/form: white pwdr. Strem# 07-0284. http://www.strem.com/catalog/v/07-0284/
Catalog No. 50-901-08924 Supplier Strem, An Ascensus Company Supplier No. 0702841G
CAS# 851477-20-8. 1g. 1-[3,5-Bis(trifluoromethyl)phenyl]-3-[(1S,2S)-(+)-2-(dimethylamino)cyclohexyl]thiourea (S,S-TUC). MFCD09834840. Molecular Weight: 413.42. Molecular Formula: C17H21F6N3S. Color/form: white pwdr. Strem# 07-0284. http://www.strem.com/catalog/v/07-0284/