Learn More
Medchemexpress LLC 1-[3,5-bis(trifluoromethyl)benzoyl]-N-(4-chlorophenyl)-3-piperidinecarboxamide | 1207113-88-9 | 99.3% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY12085510MG
Encompass_Preferred
Small-molecule inhibitor of myocardin-related transcription factor A/serum response factor (MRTF-A/SRF) signaling used in research to block MRTF-A nuclear localization and probe Rho/MKL1-SRF transcriptional pathways.
  • Specific inhibitor of MRTF-A/SRF signaling for cell and molecular studies.
  • High purity suitable for research applications.
  • Molecular weight 478.82 g/mol; formula C21H17ClF6N2O2.
  • Supplied in small mg quantities for experimental use.
  • Used to study fibrosis, cytoskeletal regulation, and transcriptional pathways.
Catalog No. 50-002-30844