Learn More
Medchemexpress LLC 1-[3,5-bis(trifluoromethyl)benzoyl]-N-(4-chlorophenyl)-3-piperidinecarboxamide | 1207113-88-9 | 99.3% | 50 MG
SDP
Catalog No. 5000232148
Encompass_Preferred
CCG-100602 is a research-grade small-molecule inhibitor of myocardin-related transcription factor A/serum response factor (MRTF-A/SRF) signaling, supplied as a solid for laboratory research applications.
  • Inhibits MRTF-A/SRF signaling and reduces MRTF-A nuclear localization.
  • High analytical purity suitable for research use.
  • Available in small-scale packages for in-lab assays and dose-response studies.
  • Solid form with storage recommendations to protect from light; in solvent store at -80°C for long-term stability.
Catalog No. 50-002-32148 Supplier Medchemexpress LLC Supplier No. HY12085550MG
CCG-100602 is a research-grade small-molecule inhibitor of myocardin-related transcription factor A/serum response factor (MRTF-A/SRF) signaling, supplied as a solid for laboratory research applications.
  • Inhibits MRTF-A/SRF signaling and reduces MRTF-A nuclear localization.
  • High analytical purity suitable for research use.
  • Available in small-scale packages for in-lab assays and dose-response studies.
  • Solid form with storage recommendations to protect from light; in solvent store at -80°C for long-term stability.