Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Cyclohexanetricarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. C20295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20295G 5 g
C202925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C20295G Supplier TCI America Supplier No. C20295G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 25357-95-3
Molecular Formula C9H12O6
Molecular Weight (g/mol) 216.189
MDL Number MFCD00134148
InChI Key FTHDNRBKSLBLDA-UHFFFAOYSA-N
PubChem CID 349732
IUPAC Name cyclohexane-1,3,5-tricarboxylic acid
SMILES C1C(CC(CC1C(=O)O)C(=O)O)C(=O)O

Specifications

Melting Point 207°C
Color White
Quantity 5 g
Formula Weight 216.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Cyclohexanetricarboxylic Acid (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.