Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(3,5-dichlorophenyl)sulfonyl]-N-[(4-fluorophenyl)methyl]-4-piperidinecarboxamide | 2227549-00-8 | >98.0% | 445.34 Da | C19H19Cl2FN2O3S | 50MG
SDP

Catalog No. 5000235473
Encompass_Preferred

A small-molecule chemical probe that potently and selectively inhibits the sodium-coupled citrate transporter SLC13A5 (NaCT). It is reported to act as a high-affinity, selective, and functionally irreversible inhibitor in human cells and is used as a research tool to study citrate transport and liver metabolism.

  • Potent, selective inhibition of human SLC13A5 (IC50 = 56 nM in HEK cells; 24 nM in HepG2 cells).
  • High selectivity over other SLC13 family members (IC50 > 100 μM).
  • Chemical formula C19H19Cl2FN2O3S and molecular weight 445.34 Da.
  • Purity ≥98.0% (HPLC).
  • Low aqueous solubility (<1 μg/mL at pH 6.8); soluble in DMSO.
  • High plasma protein binding in human plasma (~99%).
  • Moderate microsomal stability and variable hepatocyte stability across species.

Catalog No. 50-002-35473 Supplier Medchemexpress LLC Supplier No. HY12473850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule chemical probe that potently and selectively inhibits the sodium-coupled citrate transporter SLC13A5 (NaCT). It is reported to act as a high-affinity, selective, and functionally irreversible inhibitor in human cells and is used as a research tool to study citrate transport and liver metabolism.

  • Potent, selective inhibition of human SLC13A5 (IC50 = 56 nM in HEK cells; 24 nM in HepG2 cells).
  • High selectivity over other SLC13 family members (IC50 > 100 μM).
  • Chemical formula C19H19Cl2FN2O3S and molecular weight 445.34 Da.
  • Purity ≥98.0% (HPLC).
  • Low aqueous solubility (<1 μg/mL at pH 6.8); soluble in DMSO.
  • High plasma protein binding in human plasma (~99%).
  • Moderate microsomal stability and variable hepatocyte stability across species.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.