Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-((3,5-Difluorophenyl)Sulfonyl)Bicyclo[2.1.0]Pentane

Catalog No. ALD005602 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ALD005602 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ALD005602 Supplier Sigma Aldrich Supplier No. ALD00560250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The following spring-loaded electrophile is a convenient reagent for the installation of cyclopentane to primary and secondary amines. This product is one of several synthetically useful reagents reported by Baran and coworkers on Strain-release amination.

Specifications

CAS 6305-38-0
Molecular Formula C13H17NO3S
Quantity 250 mg
Molecular Weight (g/mol) 267.34 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.