Learn More
Medchemexpress LLC 1-(3,5-dimethylphenyl)-2-[2-(trifluoromethyl)phenyl]guanidine | 1430208-73-3 | 99.7% | 307.31 g·mol⁻¹ | C16H16F3N3 | 25 MG
SDP
Catalog No. 5000241908
Encompass_Preferred
1A-116 is a small-molecule Rac1 inhibitor that selectively targets the W56 residue to prevent EGF-induced Rac1 activation and disrupt Rac1-P-Rex1 interaction. It induces apoptosis and inhibits cell proliferation, migration, and cell-cycle progression in a concentration-dependent manner, and has demonstrated antimetastatic activity in vivo. Molecular formula C16H16F3N3; molecular weight 307.31 g·mol⁻¹; reported purity 99.72%.
  • Selective Rac1 inhibition via W56 targeting.
  • Prevents EGF-induced Rac1 activation.
  • Blocks Rac1-P-Rex1 interaction.
  • Induces apoptosis and inhibits cell proliferation and migration.
  • Demonstrates antimetastatic activity in vivo.
Catalog No. 50-002-41908 Supplier Medchemexpress LLC Supplier No. HY10406425MG
1A-116 is a small-molecule Rac1 inhibitor that selectively targets the W56 residue to prevent EGF-induced Rac1 activation and disrupt Rac1-P-Rex1 interaction. It induces apoptosis and inhibits cell proliferation, migration, and cell-cycle progression in a concentration-dependent manner, and has demonstrated antimetastatic activity in vivo. Molecular formula C16H16F3N3; molecular weight 307.31 g·mol⁻¹; reported purity 99.72%.
  • Selective Rac1 inhibition via W56 targeting.
  • Prevents EGF-induced Rac1 activation.
  • Blocks Rac1-P-Rex1 interaction.
  • Induces apoptosis and inhibits cell proliferation and migration.
  • Demonstrates antimetastatic activity in vivo.