Learn More
Medchemexpress LLC 1-(3,5-dimethylphenyl)-2-[2-(trifluoromethyl)phenyl]guanidine | 1430208-73-3 | MFCD31619254 | 99.7% | 307.31 g·mol⁻¹ | C16H16F3N3 | 50 MG
SDP
Catalog No. 5000243461
Encompass_Preferred
1A-116 is a small-molecule Rac1 inhibitor for research use that blocks Rac1 activation and disrupts Rac1-P-Rex1 interaction. It prevents EGF-induced Rac1 activation and has been shown to induce apoptosis in a circadian-dependent manner. Supplied as a white to off-white solid, the compound has a molecular weight of 307.31 g·mol⁻¹ and high purity (~99.7%), appropriate for biochemical and cell-based studies.
  • Blocks Rac1 activation and Rac1-P-Rex1 interaction.
  • Prevents EGF-induced Rac1 activation in cell assays.
  • Induces apoptosis in a circadian-dependent manner in studied models.
  • High purity suitable for biochemical and cell-based experiments.
  • Available in multiple small-scale packaging options for laboratory research.
  • Stable as a powder under recommended storage conditions.
Catalog No. 50-002-43461 Supplier Medchemexpress LLC Supplier No. HY10406450MG
1A-116 is a small-molecule Rac1 inhibitor for research use that blocks Rac1 activation and disrupts Rac1-P-Rex1 interaction. It prevents EGF-induced Rac1 activation and has been shown to induce apoptosis in a circadian-dependent manner. Supplied as a white to off-white solid, the compound has a molecular weight of 307.31 g·mol⁻¹ and high purity (~99.7%), appropriate for biochemical and cell-based studies.
  • Blocks Rac1 activation and Rac1-P-Rex1 interaction.
  • Prevents EGF-induced Rac1 activation in cell assays.
  • Induces apoptosis in a circadian-dependent manner in studied models.
  • High purity suitable for biochemical and cell-based experiments.
  • Available in multiple small-scale packaging options for laboratory research.
  • Stable as a powder under recommended storage conditions.