Learn More
Medchemexpress LLC 1-(3,5-dimethylphenyl)-3-(2-(trifluoromethyl)phenyl)guanidine | 1430208-73-3 | MFCD31619254 | 99.7% | 307.31 | C16H16F3N3 | 100 MG
SDP
Catalog No. 5000242485
Encompass_Preferred
A small-molecule Rac1 inhibitor that prevents EGF-induced Rac1 activation and disrupts Rac1-P-Rex1 interaction. It induces apoptosis and inhibits cell proliferation, migration, and cell-cycle progression in a concentration-dependent manner, and has demonstrated antimetastatic activity in vivo. Supplied as a white to off-white solid, it is intended for research applications.
  • Inhibits Rac1 activation and Rac1-P-Rex1 interaction.
  • Induces apoptosis and inhibits cell proliferation, migration, and cell-cycle progression.
  • Demonstrates antimetastatic activity in vivo.
  • Supplied as a white to off-white solid and as a DMSO solution.
  • High purity suitable for research applications.
  • Compatible with in vitro and in vivo assays for Rac1 pathway studies.
Catalog No. 50-002-42485 Supplier Medchemexpress LLC Supplier No. HY104064100MG
A small-molecule Rac1 inhibitor that prevents EGF-induced Rac1 activation and disrupts Rac1-P-Rex1 interaction. It induces apoptosis and inhibits cell proliferation, migration, and cell-cycle progression in a concentration-dependent manner, and has demonstrated antimetastatic activity in vivo. Supplied as a white to off-white solid, it is intended for research applications.
  • Inhibits Rac1 activation and Rac1-P-Rex1 interaction.
  • Induces apoptosis and inhibits cell proliferation, migration, and cell-cycle progression.
  • Demonstrates antimetastatic activity in vivo.
  • Supplied as a white to off-white solid and as a DMSO solution.
  • High purity suitable for research applications.
  • Compatible with in vitro and in vivo assays for Rac1 pathway studies.