Learn More
Medchemexpress LLC 1,3,5-Tri-O-benzoyl-a-D-ribofuranose | 22224-41-5 | 99.8% | 462.45 | 50 G
SDP
Catalog No. 5000454722
Encompass_Preferred
1,3,5-Tri-O-benzoyl-a-D-ribofuranose is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies.
  • Inhibition of DNA synthesis
  • Induction of apoptosis
Catalog No. 50-004-54722 Supplier Medchemexpress LLC Supplier No. HYW01372850G
1,3,5-Tri-O-benzoyl-a-D-ribofuranose is a purine nucleoside analog. Purine nucleoside analogs exhibit broad antitumor activity, specifically targeting indolent lymphoid malignancies.
  • Inhibition of DNA synthesis
  • Induction of apoptosis