Learn More
Medchemexpress LLC 1,3,5-tri-O-benzoyl-α-D-ribofuranose | 22224-41-5 | 99.8% | 462.45 | 100 G
SDP
Catalog No. 5000454145
Encompass_Preferred
1,3,5-Tri-O-benzoyl-α-D-ribofuranose is a purine nucleoside analog known for its broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis. This product is intended for research use only and is not sold to patients.
  • Targets nucleoside antimetabolite/analog pathways.
  • Involved in cell cycle/DNA damage pathways.
  • Soluble in DMSO at 100 mg/mL (216.24 mM), requiring ultrasonication.
  • Powder is stable for 3 years at -20°C and 2 years at 4°C.
  • In solvent, stable for 6 months at -80°C and 1 month at -20°C.
Catalog No. 50-004-54145 Supplier Medchemexpress LLC Supplier No. HYW013728100G
1,3,5-Tri-O-benzoyl-α-D-ribofuranose is a purine nucleoside analog known for its broad antitumor activity, particularly against indolent lymphoid malignancies. Its anticancer mechanisms involve the inhibition of DNA synthesis and the induction of apoptosis. This product is intended for research use only and is not sold to patients.
  • Targets nucleoside antimetabolite/analog pathways.
  • Involved in cell cycle/DNA damage pathways.
  • Soluble in DMSO at 100 mg/mL (216.24 mM), requiring ultrasonication.
  • Powder is stable for 3 years at -20°C and 2 years at 4°C.
  • In solvent, stable for 6 months at -80°C and 1 month at -20°C.