Learn More
Medchemexpress LLC 1,3,5-Triazine-2,4-diamine, 6-chloro-N-(ethyl-d5)-N'-(1-methylethyl)- | 163165-75-1 | 1 MG
SDP
Catalog No. 5000418694
Encompass_Preferred
Atrazine-d5 is deuterium labeled Atrazine, primarily used for controlling certain annual broadleaf and grass weeds. It inhibits photophosphorylation but typically does not cause lethality or permanent cell damage in the short term. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs, making this compound suitable for use as a tracer or an internal standard.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-18694 Supplier Medchemexpress LLC Supplier No. HYN7091S21MG
Atrazine-d5 is deuterium labeled Atrazine, primarily used for controlling certain annual broadleaf and grass weeds. It inhibits photophosphorylation but typically does not cause lethality or permanent cell damage in the short term. Deuterium substitution can impact the pharmacokinetic and metabolic profiles of drugs, making this compound suitable for use as a tracer or an internal standard.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes are incorporated for quantitation during drug development.
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs.