Learn More
Medchemexpress LLC 1,3,5-tribromobenzene-d3 | 52921-77-4 | MFCD11983526 | >=98.0% | 317.82 g/mol | C6D3Br3 | 250 MG
SDP
Catalog No. 5000282125
Encompass_Preferred
1,3,5-Tribromobenzene-d3 is the deuterium-labeled analogue of 1,3,5-tribromobenzene used as a tracer and internal standard in analytical chemistry. It is intended for quantitative applications where isotopic labeling improves detection and quantitation, and is supplied with documentation for safe handling and use.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Approx. 99 atom % D and ≥98.0% chemical purity.
  • Molecular formula C6D3Br3; molecular weight 317.82 g/mol.
  • Available in small research pack sizes for laboratory use.
  • Provided with a product datasheet and safety data sheet for handling information.
Catalog No. 50-002-82125 Supplier Medchemexpress LLC Supplier No. HYW244384250MG
1,3,5-Tribromobenzene-d3 is the deuterium-labeled analogue of 1,3,5-tribromobenzene used as a tracer and internal standard in analytical chemistry. It is intended for quantitative applications where isotopic labeling improves detection and quantitation, and is supplied with documentation for safe handling and use.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • Approx. 99 atom % D and ≥98.0% chemical purity.
  • Molecular formula C6D3Br3; molecular weight 317.82 g/mol.
  • Available in small research pack sizes for laboratory use.
  • Provided with a product datasheet and safety data sheet for handling information.