Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific 1,3,5-Tribromobenzene, 626-39-1, MFCD00000080, 100g

Catalog No. 50426926
Encompass_Preferred

Molecular Formula C6H3Br3, Purity 95+%, Molecular Weight 314.82, Melting Point 121-124°, Boiling Point 271

Catalog No. 50-426-926 Supplier Matrix Scientific Supplier No. 076150100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C6H3Br3, Purity 95+%, Molecular Weight 314.82, Melting Point 121-124°, Boiling Point 271

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.