Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trichloro-2,4,6-trifluorobenzene, 98%, Thermo Scientific™

Catalog No. AAA1686422
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1686422 100 g
AAA1686406 5 g
AAA1686414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1686422 Supplier Thermo Scientific Chemicals Supplier No. A1686422

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319-88-0
Molecular Formula C6Cl3F3
Molecular Weight (g/mol) 235.411
MDL Number MFCD00000587
InChI Key QPXZZPSKCVNHFW-UHFFFAOYSA-N
Synonym 1,3,5-trichlorotrifluorobenzene, benzene, 1,3,5-trichloro-2,4,6-trifluoro, sym-trichlorotrifluorobenzene, sym-trifluorotrichlorobenzene, 1,3,5-trifluorotrichlorobenzene, 2,4,6-trifluorotrichlorobenzene, 1,3,5-trifluoro-2,4,6-trichlorobenzene, acmc-20ap5m, 1,5-trichlorotrifluorobenzene, wln: gr cg eg bf df ff
PubChem CID 67573
IUPAC Name 1,3,5-trichloro-2,4,6-trifluorobenzene
SMILES C1(=C(C(=C(C(=C1Cl)F)Cl)F)Cl)F

Specifications

Melting Point 62°C to 64°C
Boiling Point 79°C to 80°C (12 mmHg)
Flash Point 102°C (215°F)
Quantity 100 g
Beilstein 958307
Formula Weight 235.42
Percent Purity 98%
Chemical Name or Material 1,3,5-Trichloro-2,4,6-trifluorobenzene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-273-4
RTECSNumber DC2450000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.