Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Trichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog No. AAL0213218
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0213218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0213218 Supplier Thermo Scientific Chemicals Supplier No. L0213218
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

1,3,5-Trichlorobenzene is used in metal treatments, in industrial deodorants, in cleaners for drains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,5-Trichlorobenzene is used in metal treatments, in industrial deodorants, in cleaners for drains.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-70-3
Molecular Formula C6H3Cl3
Molecular Weight (g/mol) 181.44
MDL Number MFCD00000585
InChI Key XKEFYDZQGKAQCN-UHFFFAOYSA-N
Synonym s-trichlorobenzene, benzene, 1,3,5-trichloro, sym-trichlorobenzene, 1,3,5-trichloro-benzene, unii-2hs4m0bx3c, ccris 5946, hsdb 132, 2hs4m0bx3c, tcz, 1,5-trichlorobenzene
PubChem CID 7950
ChEBI CHEBI:49916
IUPAC Name 1,3,5-trichlorobenzene
SMILES ClC1=CC(Cl)=CC(Cl)=C1

Specifications

Melting Point 62°C to 65°C
Quantity 50 g
UN Number UN3077
Beilstein 1635233
Merck Index 14,9632
Boiling Point 208°C
Solubility Information Insoluble in water.
Formula Weight 181.45
Percent Purity 98%
Chemical Name or Material 1,3,5-Trichlorobenzene
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 203-608-6
RTECSNumber DC2100100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.