Learn More
Medchemexpress LLC 1,3,5-triiodo-2-methoxybenzene | 63238-41-5 | 99.9% | 485.83 g/mol | C7H5I3O | 10 MG
SDP
Catalog No. 5000296947
Encompass_Preferred
1,3,5-Triiodo-2-methoxybenzene is a triiodinated aromatic intermediate used in the synthesis of pharmaceutical and research compounds. It is supplied as a solid, with molecular formula C7H5I3O and molecular weight 485.83 g/mol. Follow the product safety data sheet for storage and handling due to oral toxicity and irritation hazards.
  • High purity suitable for synthetic applications (HPLC 99.86%)
  • Solid form for straightforward weighing and dissolution
  • Defined storage recommendations for long-term stability
  • Provided in small research quantities for laboratory use
  • Includes SDS and COA documentation for quality and safety
Catalog No. 50-002-96947 Supplier Medchemexpress LLC Supplier No. HY2426810MG
1,3,5-Triiodo-2-methoxybenzene is a triiodinated aromatic intermediate used in the synthesis of pharmaceutical and research compounds. It is supplied as a solid, with molecular formula C7H5I3O and molecular weight 485.83 g/mol. Follow the product safety data sheet for storage and handling due to oral toxicity and irritation hazards.
  • High purity suitable for synthetic applications (HPLC 99.86%)
  • Solid form for straightforward weighing and dissolution
  • Defined storage recommendations for long-term stability
  • Provided in small research quantities for laboratory use
  • Includes SDS and COA documentation for quality and safety