Learn More
Selleck Chemical LLC 1 3 5-Trimethoxybenzene
SDP
Catalog No. 5000003769
Encompass_Preferred
1 3 5-Trimethoxybenzene (Phloroglucinol trimethyl ether Sym-trimethoxybenzene Trimethylphloroglucinol) is a potential biomarker of flavonoid intake in human It is a human xenobiotic metabolite
Catalog No. 50-000-03769 Supplier Selleck Chemical LLC Supplier No. S629425MG
1 3 5-Trimethoxybenzene (Phloroglucinol trimethyl ether Sym-trimethoxybenzene Trimethylphloroglucinol) is a potential biomarker of flavonoid intake in human It is a human xenobiotic metabolite